Learn More
Sino Biological Recombinant Human ENO3 Protein (His Tag)
Supplier:  Sino Biological 14270H07E100
Encompass_Preferred
A DNA sequence encoding the mature form of human ENO3 (AAH17249.1) (Met1-Lys434) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-9071