Learn More
Sino Biological Recombinant Human CPNE1 Protein (SUMO Tag)
Supplier:  Sino Biological 14283H21B100
Encompass_Preferred
A DNA sequence encoding the human CPNE1 (Q99829) (Met1-Ala537) was fused with the SUMO tag at the N-terminus.
Catalog No. 50-161-9077