Learn More
Sino Biological Recombinant Human GSTK1 Protein
Supplier:  Sino Biological 14291HNAE100
Encompass_Preferred
A DNA sequence encoding the mature form of human GSTK1 (Q9Y2Q3-1) (Gly2-Leu226) was expressed with a N-terminal Met.
Catalog No. 50-161-9081