Learn More
Sino Biological Recombinant Human RRM1 Protein (His & GST Tag)
Supplier:  Sino Biological 14316H20B50
Encompass_Preferred
A DNA sequence encoding the human RRM1 (P23921) (Met1-Ser792) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-9099