Learn More
Sino Biological Recombinant Human MAP1LC3A Protein (His Tag)
Supplier:  Sino Biological 14322H08E1
Encompass_Preferred
A DNA sequence encoding the mature form of human MAP1LC3A (Q9H492-1) (Met1-Phe121) was expressed with a polyhistide tag at the C-terminus.
Catalog No. 50-161-9102