Learn More
Sino Biological Recombinant Human PC4/SUB1 Protein (His Tag)
Supplier:  Sino Biological 14326H07E100
Encompass_Preferred
A DNA sequence encoding the mature form of human SUB1 (P53999) (Met1-Leu127) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-9104