Learn More
Sino Biological 14-3-3 eta / YWHAH Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 10847MM06100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human 14-3-3 eta / YWHAH (rh 14-3-3 eta / YWHAH; Catalog#10847-H09E; Q04917; Gly2-Asn246). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8159