Learn More
Sino Biological Recombinant Human CARHSP1 Protein (His Tag)
Supplier:  Sino Biological 14333H07E100
Encompass_Preferred
A DNA sequence encoding the human CARHSP1(Q9Y2V2 ) (Met1-Ser147) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9108