Learn More
Sino Biological Recombinant Human TPPP3 Protein (His Tag)
Supplier:  Sino Biological 14360H07E100
Encompass_Preferred
A DNA sequence encoding the mature form of human TPPP3 (Q9BW30) (Met1-Lys176) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-9114