Learn More
Sino Biological Recombinant Human Cytoglobin Protein (His Tag)
Supplier:  Sino Biological 14367H07E100
Encompass_Preferred
A DNA sequence encoding the human CYGB (AAH29798.1) (Met1-Pro190) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9118