Learn More
Abcam 143Nd Metal Conjugation Kit - Lightning-Link, 100UG
Catalog No. 502672011
Encompass_Preferred
Labeling in < 4 hrs with 30 secs hands-on time using 143Nd Metal Conjugation Kit - Lightning-Link ab269828. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2011 Supplier Abcam Supplier No. AB269828100UG
Labeling in < 4 hrs with 30 secs hands-on time using 143Nd Metal Conjugation Kit - Lightning-Link ab269828. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement