Learn More
Sino Biological Recombinant Human Pleckstrin Protein (His Tag)
Supplier:  Sino Biological 14401H07E100
Encompass_Preferred
A DNA sequence encoding the mature form of human PLEK (XP_004029394.1) (Met1-Lys350) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-9130