Learn More
Sino Biological Recombinant Human BCCIP Protein (His & GST Tag)
Supplier:  Sino Biological 14442H20B100
Encompass_Preferred
A DNA sequence encoding the human BCCIP (AAH09771.1) (Met1-Val314) was expressed with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-9147