Learn More
Sino Biological Recombinant Human SRFBP1 Protein (His & GST Tag)
Supplier:  Sino Biological 14461H20B100
Encompass_Preferred
A DNA sequence encoding the human SRFBP1(Q8NEF9) (Met1-Asp429) was expressed with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-9151