Learn More
Sino Biological Recombinant Human Cip4 Protein (His Tag)
Supplier:  Sino Biological 14476H08B100
Encompass_Preferred
A DNA sequence encoding the human TRIP10 Isoform 2 (NP_004231.1) (Met1-Asn545) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-9153