Learn More
Sino Biological Recombinant Human RIOK1 Protein (His & GST Tag)
Supplier:  Sino Biological 14477H20B50
Encompass_Preferred
A DNA sequence encoding the human RIOK1 (Q9BRS2) (Met1-Lys568) was expressed with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-9155