Learn More
Sino Biological Recombinant Human Optineurin Protein (His & GST Tag)
Supplier:  Sino Biological 14478H20B100
Encompass_Preferred
A DNA sequence encoding the human OPTN (NP_068815.2) (Met1-Ile577) was expressed with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-9156