Learn More
Sino Biological Recombinant Human VAMP3 Protein (His Tag)
Supplier:  Sino Biological 14481H07E100
Encompass_Preferred
A DNA sequence encoding the human VAMP3 (Q15836) (Met1-Lys77) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9158