Learn More
Sino Biological Recombinant Human ERH Protein (His Tag)
Supplier:  Sino Biological 14483H07E100
Encompass_Preferred
A DNA sequence encoding the mature form of human ERH (P84090) (Met1-Lys104) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9160