Learn More
Sino Biological Recombinant Human CD177 Protein (hFc Tag)
Supplier:  Sino Biological 14501H02H100
Encompass_Preferred
A DNA sequence encoding the human CD177 (AAH29167.1) (Met1-Gly407) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-9168