Learn More
Sino Biological Recombinant Human PAX8 Protein (His Tag)
Supplier:  Sino Biological 14505H08E100
Encompass_Preferred
A DNA sequence encoding the mature form of human PAX8 (Q06710-1) (Pro2-Ser146) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9172