Learn More
Sino Biological Recombinant Human Cofilin Protein (His Tag)
Supplier:  Sino Biological 14544H07E100
Encompass_Preferred
A DNA sequence encoding the human CFL1 (P23528) (Met1-Leu166) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9184