Learn More
Sino Biological Recombinant Human GABARAPL1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 14549H07E500
Encompass_Preferred
A DNA sequence encoding the human GABARAPL1 (Q9H0R8) (Met1-Lys117) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-9193