Learn More
Sino Biological Recombinant Human BCL2L12 Protein (GST Tag)
Supplier:  Sino Biological 14551H09E100
Encompass_Preferred
A DNA sequence encoding the human BCL2L12 (Q9HB09-1) (Pro147-Lys227) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-9194