Learn More
Sino Biological Recombinant Human LC3B Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 14555H07E100
Encompass_Preferred
A DNA sequence encoding the human MAP1LC3B (Q9GZQ8) (Met1-Val125) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9204