Learn More
Sino Biological Recombinant Human DCBLD1 Protein (His Tag)
Supplier:  Sino Biological 14558H08H50
Encompass_Preferred
A DNA sequence encoding the human DCBLD1 (Q8N8Z6-2) (Met1-Val459) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9209