Learn More
Sino Biological Recombinant Human TNFAIP8 Protein (His Tag)
Supplier:  Sino Biological 14559H07E100
Encompass_Preferred
A DNA sequence encoding the mature form of human TNFAIP8 (O95379) (Met1-Ile198) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-9210