Learn More
Sino Biological Recombinant Human 15 Lipoxygenase 2 Protein (His & GST Tag)
Supplier:  Sino Biological 14564H20B100
Encompass_Preferred
A DNA sequence encoding the human ALOX15B (AAH35217.1)( Met1-Ile676) was expressed the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-9220