Learn More
Sino Biological Recombinant Human 15 Lipoxygenase 2 Protein
Supplier:  Sino Biological 14564HNCB100
Encompass_Preferred
A DNA sequence encoding the human ALOX15B (AAH35217.1)( Met1-Ile676) was expressed and purified with two additional amino acids (Gly & Pro ) at the N-terminus.
Catalog No. 50-161-9222