Learn More
Sino Biological Recombinant Human GCAP1 Protein (His & GST Tag)
Supplier:  Sino Biological 14565H20B100
Encompass_Preferred
A DNA sequence encoding the human GUCA1A (P43080)( Met1-Gly201) was expressed the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-9224