Learn More
Sino Biological Recombinant Human GCAP1 Protein
Supplier:  Sino Biological 14565HNCB100
Encompass_Preferred
A DNA sequence encoding the human GUCA1A (P43080)(Met1-Gly201) was expressed and purified with two additional amino acids (Gly & Pro) at the N-terminus.
Catalog No. 50-161-9226