Learn More
Sino Biological Recombinant Human VILIP1 Protein
Supplier:  Sino Biological 14571HNAE100
Encompass_Preferred
A DNA sequence encoding human VSNL1 (Gly2-Lys191) was expressed with a N-terminal Met.
Catalog No. 50-161-9240