Learn More
Sino Biological Recombinant Human XTP3TPA Protein (His Tag)
Supplier:  Sino Biological 14577H07E100
Encompass_Preferred
A DNA sequence encoding the mature form of human XTP3TPA(NP_077001.1) (Met1-Thr170) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9248