Learn More
Sino Biological Recombinant Human RKIP/PEBP1 Protein
Supplier:  Sino Biological 14582HNAE100
Encompass_Preferred
A DNA sequence encoding the human PEBP1 (NP_002558.1) (Met1-Lys187) was expressed.
Catalog No. 50-161-9252