Learn More
Sino Biological Recombinant Human ARL6IP6 Protein (mFc Tag)
Supplier:  Sino Biological 14613H04H100
Encompass_Preferred
A DNA sequence encoding the human ARL6IP6 (NP_689735.1) (Met1-Ser110) was expressed with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-161-9263