Learn More
Sino Biological Recombinant Human TPM1 Protein (His Tag)
Supplier:  Sino Biological 14615H07E100
Encompass_Preferred
A DNA sequence encoding the human TPM1 (NP_000357.3) (Met1-Met284) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-9267