Learn More
Sino Biological Recombinant Human Stomatin/STOM Protein (mFc Tag)
Supplier:  Sino Biological 14624H05H100
Encompass_Preferred
A DNA sequence encoding the human STOM (AAH10703.1) (Lys55-Gly288) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-9273