Learn More
Sino Biological Recombinant Human CBR3 Protein (His Tag)
Supplier:  Sino Biological 14632H07E100
Encompass_Preferred
A DNA sequence encoding the human CBR3 (O75828) (Met1-Trp277) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9279