Learn More
Sino Biological Recombinant Human NOTCH2NL Protein (mFc Tag)
Supplier:  Sino Biological 14636H04H100
Encompass_Preferred
A DNA sequence encoding the human NOTCH2NL (XP_006711766.1) (Met1-Asn236) was expressed with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-161-9281