Learn More
Sino Biological Recombinant Human TCTP/TPT1 Protein (His Tag)
Supplier:  Sino Biological 14662H07E100
Encompass_Preferred
A DNA sequence encoding the human TPT1 (Met1-Cys172) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9291