Learn More
Sino Biological Recombinant Human Cyclophilin F Protein (His Tag)
Supplier:  Sino Biological 14689H08H100
Encompass_Preferred
A DNA sequence encoding the human PPIF (NP_005720.1) (Cys30-Ser207) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-9304