Learn More
Sino Biological Recombinant Human Syntaxin 7 Protein (mFc Tag)
Supplier:  Sino Biological 14692H04H100
Encompass_Preferred
A DNA sequence encoding the human STX7 (NP_003560.2) (Ser2-Leu238) was expressed with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-161-9305