Learn More
Sino Biological Recombinant Human Syntaxin 7 Protein (His Tag)
Supplier:  Sino Biological 14692H07H100
Encompass_Preferred
A DNA sequence encoding the human STX7 (O15400-1) (Ser2-Leu238) was expressed with an N-terminal polyhistidine tag.
Catalog No. 50-161-9307