Learn More
Sino Biological Recombinant Human PPT1 Protein (His Tag)
Supplier:  Sino Biological 14703H08H100
Encompass_Preferred
A DNA sequence encoding the human PPT1 (P50897-1)(Met1-Gly306) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-9311