Learn More
Sino Biological Recombinant Human ABO Protein (hFc Tag)
Supplier:  Sino Biological 14734H01H100
Encompass_Preferred
A DNA sequence encoding the human ABO (ADX99264.1) (Arg63-Pro354) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-9315