Learn More
Sino Biological Recombinant Human Sphingomyelin Synthase 2 Protein (mFc Tag)
Supplier:  Sino Biological 14738H04H100
Encompass_Preferred
A DNA sequence encoding the human SGMS2 (NP_689834.1) (Met1-Thr79) was expressed with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-161-9319