Learn More
Sino Biological Recombinant Human B3GNT1 Protein (His Tag)
Supplier:  Sino Biological 14779H07H100
Encompass_Preferred
A DNA sequence encoding the human B3GNT1 (NP_006867.1) (Asp43-Cys415) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9321