Learn More
Sino Biological Recombinant Human EPHX2 Protein (His Tag)
Supplier:  Sino Biological 14856H07B100
Encompass_Preferred
A DNA sequence encoding the human EPHX2 (NP_001970.2) (Met1-Met555) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-9335