Learn More
Sino Biological Recombinant Human DPYS Protein (His & GST Tag)
Supplier:  Sino Biological 14884H20B100
Encompass_Preferred
A DNA sequence encoding the human DPYS (NP_001376.1) (Met1-Pro519) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-9337