Learn More
Sino Biological Recombinant Human DPYS Protein
Supplier:  Sino Biological 14884HNCB100
Encompass_Preferred
A DNA sequence encoding the human DPYS (NP_001376.1) (Met1-Pro519) was fused with two additional amino acids (Gly&Pro) at the N-terminus.
Catalog No. 50-161-9339